Assay Details
Gene Symbol: mcrip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044420.1 | NP_001037885.1 | 1 - 2 | 94 | 69 | |
XM_012970504.2 | XP_012825958.1 | 1 - 2 | 167 | 69 | ||
GenBank mRNA | BC159062.1 | - | 1 - 2 | 114 | 69 | |
CR760130.2 | - | 1 - 2 | 94 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
