Assay Details
Gene Symbol: mcrip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044420.1 | NP_001037885.1 | 2 - 3 | 187 | 93 | |
GenBank mRNA | BC159062.1 | - | 2 - 3 | 207 | 93 | |
CR760130.2 | - | 2 - 3 | 187 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
