Assay Details
Gene Symbol: arl5c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044444.1 | NP_001037909.1 | 3 - 4 | 297 | 72 | |
GenBank mRNA | CR760784.2 | - | 3 - 4 | 297 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
