Assay Details
| Gene Symbol: foxr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044453.1 | NP_001037918.1 | 668 | 77 | ||
| GenBank mRNA | BC122030.1 | - | 840 | 77 | ||
| CR761025.2 | - | 668 | 77 | |||
More Information
Set Membership: |
|

foxr1
Genomic Map