Assay Details
Gene Symbol: racgap1.2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044482.1 | NP_001037947.1 | 3 - 4 | 378 | 81 | |
GenBank mRNA | BC135648.1 | - | 3 - 4 | 289 | 81 | |
CR761479.2 | - | 3 - 4 | 378 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
