Assay Details
| Gene Symbol: junb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044490.1 | NP_001037955.1 | 1 | 952 | 69 | |
| GenBank mRNA | CR760158.2 | - | 1 | 952 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

junb| Gene Symbol: junb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044490.1 | NP_001037955.1 | 1 | 952 | 69 | |
| GenBank mRNA | CR760158.2 | - | 1 | 952 | 69 | |
Set Membership: |
|