Assay Details
| Gene Symbol: fyttd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044492.1 | NP_001037957.1 | 5 - 6 | 584 | 80 | |
| XM_012963095.2 | XP_012818549.1 | 5 - 6 | 693 | 80 | ||
| XM_018093927.1 | XP_017949416.1 | 4 - 5 | 577 | 80 | ||
| GenBank mRNA | BC169138.1 | - | 5 - 6 | 579 | 80 | |
| CR760495.2 | - | 5 - 6 | 584 | 80 | ||
More Information
Set Membership: |
|

fyttd1
Genomic Map