Assay Details
Gene Symbol: gemin8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045567.1 | NP_001039032.1 | 3 - 4 | 644 | 67 | |
XM_012957131.2 | XP_012812585.1 | 3 - 4 | 671 | 67 | ||
XM_012957132.1 | XP_012812586.1 | 3 - 4 | 690 | 67 | ||
XM_012957133.1 | XP_012812587.1 | 3 - 4 | 745 | 67 | ||
GenBank mRNA | BC081327.1 | - | 3 - 4 | 640 | 67 | |
BC158967.1 | - | 3 - 4 | 750 | 67 | ||
BC166061.1 | - | 3 - 4 | 601 | 67 | ||
CR762163.2 | - | 3 - 4 | 644 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
