Assay Details
Gene Symbol: rhno1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045604.1 | NP_001039069.1 | 2 - 3 | 327 | 68 | |
XM_012959175.2 | XP_012814629.1 | 2 - 3 | 532 | 68 | ||
XM_012959176.2 | XP_012814630.1 | 2 - 3 | 475 | 68 | ||
XM_012959178.2 | XP_012814632.1 | 2 - 3 | 489 | 68 | ||
XM_012959179.2 | XP_012814633.1 | 2 - 3 | 291 | 68 | ||
GenBank mRNA | BC155012.1 | - | 2 - 3 | 279 | 68 | |
CR855573.2 | - | 2 - 3 | 327 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
