Assay Details
| Gene Symbol: mzt2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045607.1 | NP_001039072.1 | 1 - 2 | 232 | 97 | |
| GenBank mRNA | BC155371.1 | - | 1 - 2 | 241 | 97 | |
| CR855602.2 | - | 1 - 2 | 232 | 97 | ||
More Information
Set Membership: |
|

mzt2b
Genomic Map