Assay Details
| Gene Symbol: coa5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045633.1 | NP_001039098.1 | 1 - 2 | 110 | 77 | |
| GenBank mRNA | CR942380.2 | - | 1 - 2 | 110 | 77 | |
More Information
Set Membership: |
|

coa5
Genomic Map