Assay Details
| Gene Symbol: hddc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045634.1 | NP_001039099.1 | 3 - 4 | 237 | 77 | |
| XM_012968545.2 | XP_012823999.1 | 3 - 4 | 354 | 77 | ||
| GenBank mRNA | BC167936.1 | - | 3 - 4 | 518 | 77 | |
| CR942384.2 | - | 3 - 4 | 237 | 77 | ||
More Information
Set Membership: |
|

hddc3
Genomic Map