Assay Details
| Gene Symbol: aes | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045637.1 | NP_001039102.1 | 3 - 4 | 462 | 88 | |
| XM_012964320.2 | XP_012819774.1 | 3 - 4 | 543 | 88 | ||
| XM_018094049.1 | XP_017949538.1 | 3 - 4 | 551 | 88 | ||
| XM_018094052.1 | XP_017949541.1 | 3 - 4 | 550 | 88 | ||
| XM_018094056.1 | XP_017949545.1 | 2 - 3 | 457 | 88 | ||
| GenBank mRNA | BC118730.1 | - | 3 - 4 | 480 | 88 | |
| BC135945.1 | - | 3 - 4 | 445 | 88 | ||
| CR942389.2 | - | 3 - 4 | 462 | 88 | ||
| GU014561.1 | - | 3 - 4 | 485 | 88 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_018094056.1 transcripts.
Set Membership: |
|

aes
Genomic Map