Assay Details
| Gene Symbol: bmp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045671.1 | NP_001039136.1 | 6 - 7 | 828 | 63 | |
| XM_012959185.2 | XP_012814639.1 | 6 - 7 | 920 | 63 | ||
| GenBank mRNA | CR942509.2 | - | 6 - 7 | 864 | 63 | |
More Information
Set Membership: |
|

bmp1
Genomic Map