Assay Details
Gene Symbol: siva1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045673.1 | NP_001039138.1 | 1 - 2 | 131 | 62 | |
GenBank mRNA | CR942512.2 | - | 1 - 2 | 131 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
