Assay Details
| Gene Symbol: wbp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045707.1 | NP_001039172.1 | 10 - 11 | 967 | 75 | |
| XM_012957171.1 | XP_012812625.1 | 10 - 11 | 1034 | 75 | ||
| XM_012957173.2 | XP_012812627.1 | 10 - 11 | 1088 | 75 | ||
| XM_012957174.2 | XP_012812628.1 | 10 - 11 | 1032 | 75 | ||
| XM_012957175.1 | XP_012812629.1 | 10 - 11 | 1065 | 75 | ||
| GenBank mRNA | BC135946.1 | - | 10 - 11 | 972 | 75 | |
| CR761869.2 | - | 10 - 11 | 967 | 75 | ||
More Information
Set Membership: |
|

wbp4
Genomic Map