Assay Details
| Gene Symbol: gramd2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045716.1 | NP_001039181.1 | 8 - 9 | 732 | 76 | |
| XM_018094110.1 | XP_017949599.1 | 8 - 9 | 1226 | 76 | ||
| GenBank mRNA | CR848441.2 | - | 8 - 9 | 732 | 76 | |
More Information
Set Membership: |
|

gramd2b
Genomic Map