Assay Details
| Gene Symbol: slc19a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045736.1 | NP_001039201.1 | 1 - 2 | 214 | 89 | |
| GenBank mRNA | CR942394.2 | - | 1 - 2 | 214 | 89 | |
More Information
Set Membership: |
|

slc19a2
Genomic Map