Assay Details
Gene Symbol: mogat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045737.1 | NP_001039202.1 | 3 - 4 | 364 | 95 | |
XM_012970512.2 | XP_012825966.1 | 3 - 4 | 427 | 95 | ||
GenBank mRNA | CR942398.2 | - | 3 - 4 | 364 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
