Assay Details
| Gene Symbol: hsp90b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045763.1 | NP_001039228.1 | 1 - 2 | 107 | 65 | |
| XM_012959200.1 | XP_012814654.1 | 1 - 2 | 165 | 65 | ||
| GenBank mRNA | BC121250.1 | - | 1 - 2 | 149 | 65 | |
| CR942671.2 | - | 1 - 2 | 107 | 65 | ||
More Information
Set Membership: |
|

hsp90b1
Genomic Map