Assay Details
| Gene Symbol: ndufa5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045800.1 | NP_001039265.1 | 3 - 4 | 237 | 82 | |
| XM_012959205.2 | XP_012814659.1 | 3 - 4 | 331 | 82 | ||
| GenBank mRNA | CR942742.2 | - | 3 - 4 | 237 | 82 | |
More Information
Set Membership: |
|

ndufa5
Genomic Map