Assay Details
Gene Symbol: rpl7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078691.1 | NP_001072159.1 | 2 - 3 | 273 | 75 | |
GenBank mRNA | BC076693.1 | - | 2 - 3 | 265 | 75 | |
BC118908.1 | - | 2 - 3 | 273 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
