Assay Details
Gene Symbol: f8a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078703.1 | NP_001072171.1 | 6 - 7 | 590 | 70 | |
GenBank mRNA | BC091084.1 | - | 6 - 7 | 520 | 70 | |
BC122015.1 | - | 6 - 7 | 590 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
