Assay Details
| Gene Symbol: shroom2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078710.1 | NP_001072178.1 | 4 - 5 | 3104 | 67 | |
| XM_012957215.2 | XP_012812669.1 | 4 - 5 | 2880 | 67 | ||
| XM_018091331.1 | XP_017946820.1 | 3 - 4 | 3790 | 67 | ||
| XM_018091332.1 | XP_017946821.1 | 3 - 4 | 2775 | 67 | ||
| GenBank mRNA | DQ886532.1 | - | 4 - 5 | 3104 | 67 | |
More Information
Set Membership: |
|

shroom2
Genomic Map