Assay Details
| Gene Symbol: tspan5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078730.1 | NP_001072198.1 | 3 - 4 | 365 | 75 | |
| XM_012964864.2 | XP_012820318.1 | 3 - 4 | 775 | 75 | ||
| GenBank mRNA | BC118743.1 | - | 3 - 4 | 365 | 75 | |
| CU075319.1 | - | 3 - 4 | 669 | 75 | ||
More Information
Set Membership: |
|

tspan5
Genomic Map