Assay Details
| Gene Symbol: galk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078731.1 | NP_001072199.1 | 7 - 8 | 1113 | 76 | |
| XM_012952544.2 | XP_012807998.1 | 8 - 9 | 1352 | 76 | ||
| GenBank mRNA | BC118744.1 | - | 7 - 8 | 1113 | 76 | |
| CU024921.1 | - | 7 - 8 | 1108 | 76 | ||
More Information
Set Membership: |
|

galk1
Genomic Map