Assay Details
| Gene Symbol: akr1c2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078736.1 | NP_001072204.1 | 4 - 5 | 468 | 64 | |
| XM_012959209.2 | XP_012814663.1 | 3 - 4 | 385 | 64 | ||
| GenBank mRNA | BC118758.1 | - | 4 - 5 | 468 | 64 | |
| CU025147.1 | - | 4 - 5 | 468 | 64 | ||
More Information
Set Membership: |
|

akr1c2
Genomic Map