Assay Details
Gene Symbol: cacng8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078749.1 | NP_001072217.2 | 3 - 4 | 788 | 68 | |
GenBank mRNA | BC118795.1 | - | 3 - 4 | 788 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
