Assay Details
Gene Symbol: cacng8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078749.1 | NP_001072217.2 | 4 - 5 | 929 | 107 | |
GenBank mRNA | BC118795.1 | - | 4 - 5 | 929 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
