Assay Details
Gene Symbol: trnau1ap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078752.1 | NP_001072220.1 | 3 - 4 | 412 | 67 | |
XM_012966728.2 | XP_012822182.1 | 4 - 5 | 358 | 67 | ||
XM_012966729.2 | XP_012822183.1 | 4 - 5 | 358 | 67 | ||
GenBank mRNA | BC118799.1 | - | 3 - 4 | 412 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
