Assay Details
Gene Symbol: rabac1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078757.1 | NP_001072225.1 | 2 - 3 | 963 | 107 | |
GenBank mRNA | BC118806.1 | - | 2 - 3 | 963 | 107 | |
BC135390.1 | - | 3 - 4 | 342 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
