Assay Details
Gene Symbol: pdss1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078761.1 | NP_001072229.1 | 7 - 8 | 760 | 77 | |
XM_012964881.2 | XP_012820335.1 | 7 - 8 | 911 | 77 | ||
GenBank mRNA | BC118813.1 | - | 7 - 8 | 760 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
