Assay Details
| Gene Symbol: frmd7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018096974.1 | XP_017952463.1 | 7 - 8 | 890 | 91 | |
| GenBank mRNA | BC118833.1 | - | 7 - 8 | 644 | 91 | |
More Information
Set Membership: |
|

frmd7
Genomic Map