Assay Details
| Gene Symbol: dhrs7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078778.1 | NP_001072246.1 | 6 - 7 | 793 | 80 | |
| XM_012970534.2 | XP_012825988.1 | 5 - 6 | 1107 | 80 | ||
| XM_012970535.2 | XP_012825989.1 | 6 - 7 | 803 | 80 | ||
| XM_012970536.2 | XP_012825990.1 | 6 - 7 | 781 | 80 | ||
| GenBank mRNA | BC118856.1 | - | 6 - 7 | 793 | 80 | |
More Information
Set Membership: |
|

dhrs7b
Genomic Map