Assay Details
Gene Symbol: smyd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078820.1 | NP_001072288.1 | 3 - 4 | 483 | 66 | |
XM_012957236.2 | XP_012812690.1 | 3 - 4 | 664 | 66 | ||
XM_012957238.2 | XP_012812692.1 | 3 - 4 | 494 | 66 | ||
XM_012957240.2 | XP_012812694.1 | 2 - 3 | 240 | 66 | ||
GenBank mRNA | BC121314.1 | - | 3 - 4 | 483 | 66 | |
CT025386.2 | - | 3 - 4 | 331 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
