Assay Details
| Gene Symbol: stmn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078859.1 | NP_001072327.1 | 220 | 77 | ||
| GenBank mRNA | BC121398.1 | - | 220 | 77 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

stmn3| Gene Symbol: stmn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078859.1 | NP_001072327.1 | 220 | 77 | ||
| GenBank mRNA | BC121398.1 | - | 220 | 77 | ||
Set Membership: |
|