Assay Details
| Gene Symbol: asna1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078873.1 | NP_001072341.1 | 469 | 84 | ||
| XM_012965665.2 | XP_012821119.1 | 3 - 4 | 646 | 84 | ||
| GenBank mRNA | BC121423.1 | - | 469 | 84 | ||
More Information
Set Membership: |
|

asna1
Genomic Map