Assay Details
| Gene Symbol: ajap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078895.1 | NP_001072363.1 | 4 - 5 | 1593 | 68 | |
| GenBank mRNA | BC121464.1 | - | 4 - 5 | 1593 | 68 | |
More Information
Set Membership: |
|

ajap1
Genomic Map