Assay Details
| Gene Symbol: spaca9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078914.2 | NP_001072382.2 | 2 - 3 | 203 | 67 | |
| XM_012968609.2 | XP_012824063.1 | 2 - 3 | 246 | 67 | ||
| GenBank mRNA | BC121496.1 | - | 2 - 3 | 67 | ||
| CX886707.1 | - | 2 - 3 | 203 | 67 | ||
More Information
Set Membership: |
|

spaca9
Genomic Map