Assay Details
| Gene Symbol: mrpl42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078932.1 | NP_001072400.1 | 3 - 4 | 172 | 68 | |
| GenBank mRNA | BC121603.1 | - | 3 - 4 | 172 | 68 | |
| CT027939.1 | - | 3 - 4 | 258 | 68 | ||
More Information
Set Membership: |
|

mrpl42
Genomic Map