Assay Details
| Gene Symbol: gpank1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078939.1 | NP_001072407.1 | 2 - 3 | 663 | 90 | |
| XM_012954978.2 | XP_012810432.1 | 2 - 3 | 842 | 90 | ||
| XM_012954979.2 | XP_012810433.1 | 2 - 3 | 836 | 90 | ||
| GenBank mRNA | BC121623.1 | - | 2 - 3 | 663 | 90 | |
More Information
Set Membership: |
|

gpank1
Genomic Map