Assay Details
Gene Symbol: rpp40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078949.1 | NP_001072417.1 | 6 - 7 | 750 | 75 | |
GenBank mRNA | BC121647.1 | - | 6 - 7 | 750 | 75 | |
CU025194.1 | - | 6 - 7 | 754 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
