Assay Details
Gene Symbol: hscb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078954.2 | NP_001072422.2 | 1 - 2 | 486 | 76 | |
XM_018094444.1 | XP_017949933.1 | 1 - 2 | 470 | 76 | ||
GenBank mRNA | BC121661.1 | - | 1 - 2 | 220 | 76 | |
CR567009.1 | - | 1 - 2 | 486 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
