Assay Details
Gene Symbol: gmeb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078964.1 | NP_001072432.1 | 7 - 8 | 891 | 69 | |
XM_012957263.2 | XP_012812717.1 | 8 - 9 | 982 | 69 | ||
XM_012957264.2 | XP_012812718.1 | 8 - 9 | 1110 | 69 | ||
XM_012957267.2 | XP_012812721.1 | 8 - 9 | 922 | 69 | ||
XM_018091341.1 | XP_017946830.1 | 9 - 10 | 1030 | 69 | ||
GenBank mRNA | BC121678.1 | - | 7 - 8 | 891 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
