Assay Details
| Gene Symbol: spty2d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078966.1 | NP_001072434.1 | 2278 | 68 | ||
| GenBank mRNA | BC121680.1 | - | 2278 | 68 | ||
| BC167401.1 | - | 2278 | 68 | |||
More Information
Set Membership: |
|

spty2d1
Genomic Map