Assay Details
Gene Symbol: gba3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078970.1 | NP_001072438.1 | 2 - 3 | 232 | 56 | |
XM_012965261.2 | XP_012820715.1 | 2 - 3 | 194 | 56 | ||
XM_018094457.1 | XP_017949946.1 | 2 - 3 | 220 | 56 | ||
GenBank mRNA | BC121688.1 | - | 2 - 3 | 232 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
