Assay Details
Gene Symbol: tspan16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078971.1 | NP_001072439.1 | 2 - 3 | 117 | 72 | |
GenBank mRNA | BC121689.1 | - | 2 - 3 | 117 | 72 | |
BC157239.1 | - | 2 - 3 | 98 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
