Assay Details
Gene Symbol: nat16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078986.1 | NP_001072454.1 | 2 - 3 | 422 | 63 | |
XM_012953374.2 | XP_012808828.1 | 2 - 3 | 415 | 63 | ||
XM_012953375.2 | XP_012808829.1 | 3 - 4 | 527 | 63 | ||
GenBank mRNA | BC121852.1 | - | 2 - 3 | 422 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
