Assay Details
| Gene Symbol: dnaaf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079006.1 | NP_001072474.1 | 1 - 2 | 1749 | 90 | |
| XM_018096316.1 | XP_017951805.1 | 1 - 2 | 2178 | 90 | ||
| GenBank mRNA | BC121889.1 | - | 1 - 2 | 1749 | 90 | |
| CR942535.2 | - | 1 - 2 | 1727 | 90 | ||
More Information
Set Membership: |
|

dnaaf2
Genomic Map