Assay Details
| Gene Symbol: itprid2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079133.2 | NP_001072601.2 | 11 - 12 | 3031 | 94 | |
| XM_018097149.1 | XP_017952638.1 | 11 - 12 | 2724 | 94 | ||
| GenBank mRNA | BC122975.1 | - | 11 - 12 | 2978 | 94 | |
| DT419235.1 | - | 1 - 2 | 395 | 94 | ||
More Information
Set Membership: |
|

itprid2
Genomic Map